missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AC 5216
GREENER_CHOICE

Product Code. 18756218
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18756218

Brand: Tocris Bioscience™ 5281/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TSPO ligand

AC 5216 is a ligand of translocator protein (TSPO, peripheral benzodiazepine receptor). Exhibits high affinity for TSPO in rat whole brain (Ki = 0.297 nM), and high potency against human and rat glial TSPO (IC50 values are 2.73 and 3.04 nM respectively). Displays no noticeable binding to a number of other receptors, transporters or ion channels. Also displays anxiolytic and antidepressant effects in rodent models, without inducing benzodiazepine-like adverse effects.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type AC 5216
Purity >98%
Molecular Weight (g/mol) 401.46
Inhibitors AC 5216

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.