missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AC 265347

Product Code. 18738239
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738239 10 mg 10mg
18720138 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18738239 Supplier Tocris Bioscience™ Supplier No. 6165/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CaSR biased allosteric modulator

AC 265347 is a CaSR biased allosteric modulator (pEC50 = 7.8-8.1). Calcimimetic. Displays ability to bias signalling towards the accumulation of pERK1/2 and IP1. Reduces parathyroid hormone (PTH) levels in rat serum without inducing the release of calcitonin. Exhibits no significant activation of human GABAB or type I PTH receptors. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material α-(2,4-Dimethylphenyl)-α-methyl-2-benzothiazolemethanol
CAS 1253901-26-6
Assay Percent Range >99%
Quantity 10 mg
Molecular Formula C17H17NOS
Formula Weight Observed MW: 283.39
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.