Learn More
Karl Hecht™ AC-15 Blood Cell Counter
Product Code. 10368861
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. Quantity Unit Size
10368861 Each Each
1 options
Product Code. 10368861 Supplier Karl Hecht™ Supplier No. 40345015
  • ONLINE EXCLUSIVE PRICE
    £1123.50
    £732.60 / Pack of 1
    Save £390.90
    34% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
An electronic counter and storage device for the microscopial analysis of differential blood count.
  • For manual differential counting
  • Microscopic image may be monitored continuously while the resulting figures are recorded “blindly”
  • 10 control keys, 15 memory keys with LED display
  • Four defined programs and one free counting program: LEUKO, MYELO, RETI, RBC, FREE
  • Results in absolute values and percentage
  • With cable, safety plug and operating instructions
Specifications
Product Type Blood Cell Counter
Description 230 V, 50 Hz
Memory 15 memory keys
Quantity Each
For Use With (Application) Microscopic analysis of differential blood count
Includes Cable, safety plug, instructions