missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ABX 464

Product Code. 18756436
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18756436 10 mg 10mg
18739836 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18756436 Supplier Tocris Bioscience™ Supplier No. 6460/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HIV RNA splicing modulator

ABX 464 is an HIV RNA splicing modulator; it selectively enhances the splicing of HIV RNA in infected human PBMCs and inhibits viral replication, with no global effect on cellular splicing. ABX 464 inhibits replication of HIV strains harboring mutations associated with drug-resistance. ABX 464 binds the cap binding complex (CBC) and upregulates miR-124 expression in vitro and decreases expression of miR-124 target genes, MCP-1, CXCL-1, SERPIN-E1 in macrophages. It also decreases the number of Th17 cells, as well as IL-6 soluble receptor in CD4+ T cells. In HIV infected humanized mice, treatment with ABX 464 reduces viral load. Anti-inflammatory.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Chloro-N-[4-(trifluoromethoxy)phenyl]-2-quinolinamine
CAS 1258453-75-6
Quantity 10 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C16H10ClF3N2O
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.