missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ ABX 464

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
ABX 464 is an HIV RNA splicing modulator; it selectively enhances the splicing of HIV RNA in infected human PBMCs and inhibits viral replication, with no global effect on cellular splicing. ABX 464 inhibits replication of HIV strains harboring mutations associated with drug-resistance. ABX 464 binds the cap binding complex (CBC) and upregulates miR-124 expression in vitro and decreases expression of miR-124 target genes, MCP-1, CXCL-1, SERPIN-E1 in macrophages. It also decreases the number of Th17 cells, as well as IL-6 soluble receptor in CD4+ T cells. In HIV infected humanized mice, treatment with ABX 464 reduces viral load. Anti-inflammatory.
Specifications
Specifications
| Chemical Name or Material | 8-Chloro-N-[4-(trifluoromethoxy)phenyl]-2-quinolinamine |
| CAS | 1258453-75-6 |
| Quantity | 50 mg |
| Target | DNA, RNA and Protein Synthesis Compounds |
| Molecular Formula | C16H10ClF3N2O |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction