missing translation for 'onlineSavingsMsg'
Learn More

ABX-1431, MedChemExpress

Product Code. 16435666
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16435666

Brand: Medchem Express HY117632

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ABX-1431 is a highly potent, selective, and orally available, CNS-penetrant monoacylglycerol lipase (MAGL) inhibitor with an IC50 of 14 nM.

ABX-1431 is a highly potent, selective, and orally available, CNS-penetrant monoacylglycerol lipase (MAGL) inhibitor with an IC50 of 14 nM.

Specifications

Chemical Name or Material ABX-1431
Physical Form Powder
CAS 1446817-84-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H22F9N3O2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 125 mg/mL (246.36 mM; Need ultrasonic)
SMILES O=C(N1CCN(CC2=CC=C(C(F)(F)F)C=C2N3CCCC3)CC1)OC(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 507.39
Formula Weight 507.39
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.