missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress ABTS diammonium salt

Product Code. 16421569
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16421569 Supplier MedChemExpress Supplier No. HY15902
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ABTS diammonium salt is a substrate for horseradish peroxidase (HRP) conjugate.

Specifications

CAS 30931-67-0
Solubility H2O : ≥ 50 mg/mL (91.13 mM) ∣DMSO : 20.83 mg/mL (37.96 mM∣Need ultrasonic)
Synonym AzBTS-(NH4)2
Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Quantity 1 g
Molecular Weight (g/mol) 548.68 g/mol
Product Type Dye Reagents
Purity or Quality Grade 98.44%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.