missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22333
This item is not returnable.
View return policy
Description
ABT1 Polyclonal specifically detects ABT1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ABT1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| activator of basal transcription 1, Basal transcriptional activator, hABT1, TATA-binding protein-binding protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 29777 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg | |
| NP_037507.1 | |
| ABT1 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Activator of Basal Transcription 1 using the numbering given in entry NP_037507.1 (GeneID 29777). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction