missing translation for 'onlineSavingsMsg'
Learn More

ABT-072, MedChemExpress

Product Code. 16440456
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16440456

Brand: Medchem Express HY101634

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress ABT-072 is an orally active and potent non-nucleoside HCV NS5B polymerase inhibitor (HCV GT1a EC50=1 nM; HCV GT1b EC50=0.3 nM).

ABT-072 is an orally active and potent non-nucleoside HCV NS5B polymerase inhibitor (HCV GT1a EC50=1 nM

Specifications

Chemical Name or Material ABT-072
CAS 1132936-00-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H27N3O5S
Quantity 20 mg
Solubility Information DMSO : 80 mg/mL (170.38 mM; Need ultrasonic)
SMILES CS(=O)(NC1=CC=C(/C=C/C2=CC(N(C(N3)=O)C=CC3=O)=CC(C(C)(C)C)=C2OC)C=C1)=O
Molecular Weight (g/mol) 469.55
Formula Weight 469.55
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.