missing translation for 'onlineSavingsMsg'
Learn More

ABT-046, MedChemExpress

Product Code. 16498947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498947 5 mg 5mg
16456666 100 mg 100mg
16442477 50 mg 50mg
16406368 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16498947 Supplier Medchem Express Supplier No. HY15197
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).

ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).

Specifications

Chemical Name or Material ABT-046
CAS 1031336-60-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22N4O2
Quantity 5 mg
Solubility Information DMSO : 66.67 mg/mL (190.26 mM; Need ultrasonic)
SMILES NC1=C(C2=CC=C([C@H]3CC[C@H](CC(O)=O)CC3)C=C2)C=NC4=CC=NN41
Molecular Weight (g/mol) 350.41
Formula Weight 350.41
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.