missing translation for 'onlineSavingsMsg'
Learn More

New Pig™ Absorbent Drip Pan

Product Code. 15340915
Encompass_Preferred
Click to view available options
Capacity (English):
18 gal.
Unit Size:
Pack of 24
This item is not returnable. View return policy

Product Code. 15340915

Brand: New Pig™ 2410PP

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Certifications/Compliance CERCLA, OSHA Hazard Communication Standard, 29 CFR 1910.1200
Color Black/Gray
Capacity (English) 18 gal.
Material Polypropylene
Type Absorbent Drip Pan
For Use With (Application) Oils, Coolants, Solvents, Water
Packaging Quantity 24/Pk.
Dimensions (L x W x H) 27 x 27 x 8 cm
Length (Metric) 27 cm
Height (Metric) 8 cm
Weight (Metric) 10.3 kg
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.