missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute QPCR Mix, no ROX

Product Code. p-7184554
Click to view available options
No. of Reactions:
1600 Reactions
Unit Size:
1600 reactions
This item is not returnable. View return policy

Product Code. 11807682

Brand: Thermo Scientific™ AB1132B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific ABsolute qPCR Mixes are optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes.

Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.

  • Thermo Scientific ABsolute qPCR Mixes are optimized for use with hybridization probes
  • They contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes
  • The 2X master mix contains a proprietary reaction buffer that provides high sensitivity, specificity, and reproducibility
  • ABsolute qPCR Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, and with no ROX reference dye
  • There is also a master mix version optimized for capillary instruments.

Highlights

  • High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
  • Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
  • Optimized buffer - For maximal performance in multiplex reactions
  • Applications - Gene expression analysis, siRNA validation, and genotyping

Alternative Product

  • Try TaqMan Fast Advanced Master Mix (4444963), our highest-performance, probe-based master mix
  • TaqMan Fast Advanced Master Mix features the best of ABsolute QPCR Mix, with added additional capabilities for gene expression analysis
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 16 x 1.25 mL ABsolute QPCR Mix (2X)

Store at -20°C
Description ABsolute QPCR Mix, QPCR mix; 1.25mL vial; 800 rxns
Detection Method Primer-probe
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon
Product Line ABsolute
Product Type qPCR Mix
Quantity 16 x 1.25 mL vials; 800 x 50 μL reactions
Sufficient For 1600 Reactions
For Use With (Application) Gene Expression, Genotyping, siRNA Validation
Fidelity (vs. Taq) 2 X
No. of Reactions 1600 Reactions
Sample Type DNA, dsDNA
Volume 1.25 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.