Learn More
ABsolute ABsolute™ qPCR Master Mixes, SYBR Green-Based Detection
Product Code. 11882043
Click to view available options
Description:
ABsolute QPCR SYBR Green Fluorescein Mix
ABsolute QPCR SYBR Green Mix
ABsolute QPCR SYBR Green ROX (500nM) Mix
ABsolute qPCR SYBR Green Low ROX Mix
Quantity:
100 x 50µL rxns
200 x 50µL rxns
2000 x 50µL rxns
800 x 50µL rxn
800 x 50µL rxns
Unit Size:
1600 reactions
200 reactions
400 reactions
4000 reactions
Product Code. 11882043
Brand: ABsolute AB1163/B
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ ABsolute qPCR Master Mixes offer sensitivity and reproducibility for real-time PCR applications.
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ SYBR Green-Based Detection qPCR Master Mixes.
Highlights
  • High Sensitivity & Specificity -The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
  • Optimised buffer containing SYBR Green I
  • Reproducibility -All master mixes undergo extensive QC to ensure batch to batch consistency
  • Broad dynamic range
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, SYBR Green I, ROX™ passive reference dye (only in ROX mixes), Fluorescein (only in Fluorescein mix)); Separate MgCl2; Separate ROX vial (only in kits indicated). Store at -20°C - Avoid freeze/thaw cycles
Description ABsolute QPCR SYBR Green ROX (500nM) Mix
Quantity 2000 x 50µL rxns
Activation Time heated to 95°C for 15 minutes
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.