Learn More
Applied Biosystems™ Absolute Q™ MAP16 Plate Kit and Absolute Q™ Universal DNA dPCR Master Mix
Shop All Applied Biosystems ProductsDescription
The Absolute Q MAP16 Plate Kit and Absolute Q Universal DNA Digital PCR Master Mix are key components of the QuantStudio Absolute Q Digital PCR System. The system is powered by microfluidic array plate (MAP) technology that enables consistent and automated separation of the sample into 20,000 microchambers with industry leading <1% coefficient of variation (CV).
Absolute Q MAP16 Plate Kit
The Absolute Q MAP16 microinjection-molded plates overcome common challenges of inconsistency and high dead volume. This novel approach provides many benefits, including precise microchamber volume, consistent sample distribution, and greater than 98% of loaded volume analyzed.
Absolute Q Universal DNA Digital PCR Master Mix
This package product also includes Absolute Q Universal DNA Digital PCR Master Mix, which is compatible with a broad range of applications and over 20 million predesigned TaqMan Assays (performance-guaranteed*), reducing the need to design custom assays. Simplify your workflow by avoiding assay design and minimizing assay optimization; use the same assay for both qPCR and digital PCR. In addition, with 72 hours of assembled-reaction benchtop stability, this master mix enables increased productivity through automation.
The impressive precision and accuracy of the Absolute Q system helps provide confidence in measurements across replicates and experiments.
* We guarantee that our predesigned TaqMan Assays will enable you to obtain data you can trust, or we'll replace them at no cost or give you a credit for the assay purchase price. Terms and conditions apply.
Specifications
Specifications
| Content And Storage | 12 MAP16 Plates, Gaskets, Isolation Buffer, and 1 tube Absolute Q™ Universal DNA Digital PCR Master Mix (192 reactions) |
| Detection Method | Primer-probe |
| Form | Liquid |
| GC-Rich PCR Performance | High |
| PCR Method | Digital PCR |
| Reaction Speed | Fast or Standard |
| Quantity | 1 Ea. |
| For Use With (Application) | Digital PCR (dPCR), DNA Quantitation, Gene Expression, Genotyping, Microbial Detection |
| No. of Reactions | 192 Reactions |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.