missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Absolute Q™ 1-step RT-dPCR Master Mix (4X)

Product Code. 17846511 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
200 reactions
Unit Size:
450µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17846511 200 reactions 450µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17846511 Supplier Applied Biosystems™ Supplier No. A55146
Focus on Regenerative Therapies
Use promo code 28027
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Applied Biosystems Absolute Q 1-step RT-dPCR Master Mix (4X) is optimized for use with the QuantStudio Absolute Q Digital PCR System in a simple workflow with minimal processing steps.

  • Optimized for use with the QuantStudio Absolute Q Digital PCR System in a simple workflow with minimal processing steps
  • The 4X formulation enables analysis of higher sample volumes and delivers accurate quantification of RNA targets without using a standard curve
  • Set up the reaction like 1-Step RT-qPCR, then load and run the plate in a single instrument
  • Requires just 5 minutes of hands-on time and RT-dPCR run time of ∼65 minutes
  • Ability to detect less than 1 copy/μL of a specific target within a 4-plex reaction
  • Detect up to four targets in a single reaction
  • Highly reproducible results between technical replicates, consistently generate >20,000 analyzable micro-chambers
TRUSTED_SUSTAINABILITY

Specifications

Detection Method Primer-probe
PCR Method Digital PCR
Polymerase Taq DNA Polymerase
Reaction Speed <2 hr.
Technique Digital PCR
Quantity 200 reactions
Compatibility Absolute Q Digital PCR System
Hot Start Hot Start Taq Polymerase
No. of Reactions 200 Reactions
Sample Type RNA
Volume 450 μL
Show More Show Less

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.