Learn More
Thermo Scientific™ ABsolute QPCR Capillary Mix
Thermo Scientific™ ABsolute qPCR Master Mixes offer sensitivity and reproducibility for real-time PCR applications.
Brand: Thermo Scientific™ AB1283B
1048.05 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : LE
Additional Details : Weight : 0.08000kg
Description
Enjoy sensitivity and reproducibility in real-time PCR applications with Thermo Scientific™ ABsolute™ Probe-Based Detection qPCR Master Mixes.
ABsolute qPCR Master Mixes are designed to achieve the most consistent and reproducible data across all QPCR platforms. Proprietary additives give enhanced and consistent end point readings as well as low Ct values. All mixes contain Thermo-Start Taq DNA Polymerase, for increased specificity, and dTTP for increased reaction efficiency and sensitivity (low Ct).Maximum specificity—Chemically-modified Thermo-Start™ Taq remains completely inactive until heated to 95°C for 15 minutes, thereby preventing nonspecific amplification
Improved efficiency—The use of dTTP in place of dUTP increases reaction efficiency by approximately 5% and therefore reduces reaction time
Increased sensitivity—The combination of Thermo-Start Taq and dTTP increases sensitivity and allows for detection of low copy number templates
Instrument specific—Individually optimized qPCR mixes designed for all major qPCR / Real-Time PCR instruments
Reproducible results—All mixes undergo extensive quality control testing to ensure batch-to-batch consistency
Simple protocol—Master mix format reduces protocol steps and minimizes risk of pipetting errors and sample contamination
Includes:
ABsolute qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated)
Recommended for:
Ideal for qPCR analysis in molecular biology applications, including gene expression and SNP typing
Specifications
Contains: • 16 x 1.25 mL ABsolute QPCR Capillary Mix (2X) Store at -20°C |
|
Primer-probe | |
Taq DNA Polymerase | |
ABsolute | |
2000 x 20 μL Reactions | |
DNA, dsDNA |
ABsolute QPCR Capillary Mix, 2000 rxns | |
qPCR | |
Standard | |
qPCR Capillary Mix | |
2000 Reactions |
For Research Use Only. Not for use in diagnostic procedures.