missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ABsolute Blue QPCR Mix, with separate ROX vial
ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity.
£456.00 - £1650.00
Specifications
Detection Method | Primer-probe |
---|---|
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10657123
|
Thermo Scientific™
AB4136B |
1600 Reactions |
£1650.00
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11832153
|
Thermo Scientific™
AB4137A |
400 Reactions |
£456.00
400 reactions |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific™ ABsolute Blue qPCR Mixes contain an inert blue dye to significantly enhance the contrast between reagent and plastic, allowing quick and easy verification that master mix has been dispensed.
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with different ROX passive reference dye concentrations
Specifications
Primer-probe | |
High | |
Taq DNA Polymerase | |
BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon | |
qPCR Mix | |
2 X |
Tube | |
qPCR | |
Standard | |
ABsolute Blue | |
2X | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.