missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute Blue QPCR Mix, with separate ROX vial

Product Code. 11832153
Change view
Click to view available options
No. of Reactions:
1600 Reactions
400 Reactions
Unit Size:
400 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
11832153 400 Reactions 400 reactions
10657123 1600 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11832153 Supplier Thermo Scientific™ Supplier No. AB4137A
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity.

Thermo Scientific™ ABsolute Blue qPCR Mixes contain an inert blue dye to significantly enhance the contrast between reagent and plastic, allowing quick and easy verification that master mix has been dispensed.

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with different ROX passive reference dye concentrations
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2X
Content And Storage Contains:
• 1 x 5 mL ABsolute Blue QPCR Mix (2X)
• 100 μL MgCl2 (1M)
• 25 μL ROX Reference Dye (1 mM)

Store at -20°C
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Standard
For Use With (Equipment) BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon
Product Line ABsolute Blue
Product Type qPCR Mix
Quantity 400 reactions
Sufficient For 400 Reactions
Fidelity (vs. Taq) 2 X
No. of Reactions 400 Reactions
Sample Type DNA, dsDNA
Volume 5 mL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.