missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ABsolute Blue QPCR Mix, SYBR Green, ROX
Click to view available options
No. of Reactions:
1600 Reactions
400 Reactions
Unit Size:
1600 reactions
400 reactions
Description
Thermo Scientific ABsolute Blue qPCR SYBR Green Mixes are optimized 2X master mixes for SYBR Green chemistry. The master mixes incorporate an inert blue dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof. Absolute Blue qPCR Mixes include Thermo-Start Taq hot start enzyme for increased sensitivity.
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with ROX or Fluorescein passive reference dyes
Specifications
Specifications
| Concentration | 2X |
| Content And Storage | Contains: • 1 x 5 mL ABsolute Blue QPCR SYBR Green ROX Mix (2X) • 100 μL MgCl2 (1M) Sufficient for 400 x 25 μL reactions. Store at -20°C |
| Detection Method | SYBR |
| Format | Tube |
| GC-Rich PCR Performance | High |
| PCR Method | qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System |
| Product Line | ABsolute Blue |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction