missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ABsolute Blue qPCR Mixes

Product Code. p-4526951 Shop All Thermo Scientific Products
Click to view available options
Product Type:
QPCR Mix and ROX Vial
QPCR ROX Mix
QPCR low ROX Mix
Quantity:
2 x 1.25mL
10 x 5mL
Sufficient For:
200 x 25µL Rxns
4000 x 25µL Rxns
Unit Size:
200 reactions
4000 reactions
This item is not returnable. View return policy

Product Code. 11842153

Brand: Thermo Scientific™ AB4137/B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Make verification of master mix dispensing quick, easy and foolproof with these colored qPCR master mixes for probe chemistry using standard cycling protocols.

ABsolute Blue qPCR Mixes are 2X master mixes for probe chemistry. There are suitable alternatives available for all common real-time instruments. ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity. The master mixes incorporate an inert blue dye to enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

Highlights

  • High performance - ABsolute sensitivity, consistency & convenience
  • Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
  • Universal - compatible with all major qPCR platforms
  • Adaptable - available with different ROX passive reference dye concentrations
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ABsolute Blue qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated). Store at -20°C. Avoid freeze/thaw cycles. The ROX dye is light sensitive; exposure to light should be minimized for vials containing ROX dye.
Product Type QPCR Mix and ROX Vial
Quantity 10 x 5mL
Sufficient For 4000 x 25µL Rxns
For Use With (Application) Gene Expression
Compatibility ABsolute Blue qPCR mixes are ideal for use with white reaction plates and tubes. Check qPCR cycler compatibility with our qPCR reagent selection guide.

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.