Learn More
Thermo Scientific™ ABsolute Blue qPCR Mixes
Shop All Thermo Scientific ProductsDescription
ABsolute Blue qPCR Mixes are 2X master mixes for probe chemistry. There are suitable alternatives available for all common real-time instruments. ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity. The master mixes incorporate an inert blue dye to enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.
Highlights
- High performance - ABsolute sensitivity, consistency & convenience
- Enhanced contrast and reduced errors - easily identify correctly aliquotted reaction mix in all plates and tubes
- Universal - compatible with all major qPCR platforms
- Adaptable - available with different ROX passive reference dye concentrations
Specifications
Specifications
| Content And Storage | ABsolute Blue qPCR Buffer (Thermo-Start DNA polymerase, dNTPs, MgCl2, ROX™ passive reference dye (only in ROX mixes); Separate ROX vial (only in kits indicated). Store at -20°C. Avoid freeze/thaw cycles. The ROX dye is light sensitive; exposure to light should be minimized for vials containing ROX dye. |
| Product Type | QPCR ROX Mix |
| Quantity | 10 x 5mL |
| Sufficient For | 4000 x 25µL Rxns |
| For Use With (Application) | Gene Expression |
| Compatibility | ABsolute Blue qPCR mixes are ideal for use with white reaction plates and tubes. Check qPCR cycler compatibility with our qPCR reagent selection guide. |
Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.