Learn More
BD AbSeq Enhancer Kit
Product Code. 32188681 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
AbSeq Enhancer Kit
Unit Size:
25µg
Product Code. Product Type Unit Size
32188681 AbSeq Enhancer Kit 25µg
1 options
Product Code. 32188681 Supplier BD Supplier No. 570750
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Improves BD™ AbSeq performance by diminishing undesired Ab-Oligo background noise that may occur in certain applications.
This product should be used in workflows where samples are preserved in BD™ OMICS-Guard Sample Preservation Buffer prior to BD™ AbSeq staining, such as with BD™ Intracellular AbSeq. BD™ AbSeq Enhancer is a combination of 3 individual components that should be added in equal amounts to cells before they are stained with BD™ AbSeq.
TRUSTED_SUSTAINABILITY
Specifications
Product Type AbSeq Enhancer Kit