missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Absent in melanoma 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | Absent in melanoma 1 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18668795
|
Novus Biologicals
NBP2-38171-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179367
|
Novus Biologicals
NBP2-38171 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Absent in melanoma 1 Polyclonal specifically detects Absent in melanoma 1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Absent in melanoma 1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| AIM1, Beta/gamma crystallin domain-containing protein 1, beta-gamma crystallin domain containing 1, CRYBG1, ST4, suppression of tumorigenicity 4 (malignant melanoma) | |
| AIM1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9Y4K1 | |
| 202 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DDMEKANHIESVIKSNLPNCANSDTDFMGLFKSSRYDPSISFSGMSLSDTMTLRGSVQNKLNPRPGKVVIYSEPDVSEKCIEVFSDIQDCSSWS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title