missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AbSelect™ Serum Antibody Purification Kit
Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
Description
AbSelect Serum is prepared by coupling highly purified protein A to agarose beads which can be used to purify IgG fractions from ascites fluid and serum. The method involves the capture of the antibody on the AbSelect Serum resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
- Compatible with the lightning-link antibody labeling kits
- Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Sufficient For | 1 Purification |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 1 Test |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction