missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AbSelect™ Mouse TCS Antibody Purification Kit
Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
Description
AbSelect Mouse TCS resin has a high affinity for mouse IgG molecules. The method involves capture of the antibody on the AbSelect Mouse TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.
- This system is not suitable for use with antibodies from other species
- Binding strength of Bovine IgG to AbSelect Mouse resin is negligible
- Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Sufficient For | 3 Purifications |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 3 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction