missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AbSelect™ Mouse TCS Antibody Purification Kit

Product Code. 18721144 Shop All Bio Techne Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
missing translation for 'unitSize'
1 test
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For unitSize
18721144 1 Purification 1 test
18747713 3 Purifications 3 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 18721144 missing translation for 'mfr' Novus Biologicals™ missing translation for 'supplierNo' 8320005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For purification of mouse IgG fractions from hybridoma supernatants

AbSelect Mouse TCS resin has a high affinity for mouse IgG molecules. The method involves capture of the antibody on the AbSelect Mouse TCS resin and the removal of unwanted substances using a simple wash procedure. The purified product is then eluted and neutralized.

  • This system is not suitable for use with antibodies from other species
  • Binding strength of Bovine IgG to AbSelect Mouse resin is negligible
  • Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system

Recommended Storage

Store at 4°C. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Purification Kit
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.