missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AbSelect™ Mouse Antibody Purification Kit

Product Code. 18073494 Shop All Bio Techne Products
Change view
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sufficient For Unit Size
18073494 3 Purifications 3 tests
18023224 1 Purification 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18073494 Supplier Novus Biologicals™ Supplier No. 8300010
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For purification of mouse antibodies from crude samples such as ascites fluid

  • Mouse antibody to be purified or cleaned up is in volume of 0.1 to 0.5mL
  • Up to 150g of antibody can be purified in each run
    • This system is not suitable for use with antibodies from other species
    • Components of the AbSelect kit are fully compatible with lightning-link and lightning-link rapid antibody labeling system

Recommended Storage

Store at 4°C. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Purification Kit
Sufficient For 3 Purifications
Content And Storage 4°C. Do not freeze.
Quantity 3 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.