missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ AbSelect™ G Serum Antibody Purification Kit
Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
Description
Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species. AbSelect G Serum resin is prepared by coupling purified Protein G to agarose beads. It can therefore be used to purify IgG fractions from both serum and ascites fluid.
- Compatible with the lightning-link antibody labeling kits
- Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye, with a hands-on time of under 30 seconds
Recommended Storage
Store at 4°C. Do not freeze.
Specifications
Specifications
| Product Type | Antibody Purification Kit |
| Sufficient For | 3 Purifications |
| Content And Storage | 4°C. Do not freeze. |
| Quantity | 3 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction