missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ AbSelect™ G Serum Antibody Purification Kit

Product Code. 18065704 Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
This item is not returnable. View return policy

Product Code. 18065704

Brand: Novus Biologicals™ 8930030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For purification of antibodies from crude samples such as ascites fluid or immune serum

Protein G has a high affinity for the Fc regions of IgG molecules from a variety of species. AbSelect G Serum resin is prepared by coupling purified Protein G to agarose beads. It can therefore be used to purify IgG fractions from both serum and ascites fluid.

  • Compatible with the lightning-link antibody labeling kits
  • Allows purified antibody to be immediately labeled with an enzyme, fluorescent protein or fluorescent dye, with a hands-on time of under 30 seconds

Recommended Storage

Store at 4°C. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Purification Kit
Sufficient For 1 Purification
Content And Storage 4°C. Do not freeze.
Quantity 1 Test
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.