missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ AbSelect™ Antibody Concentration and Clean Up Antibody Purification Kit

Product Code. 18769673
Change view
Click to view available options
Quantity:
3 Tests
Unit Size:
3 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18769673 3 Tests 3 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18769673 Supplier R&D Systems™ Supplier No. 8610010
  • ONLINE EXCLUSIVE PRICE
    £351.00
    £331.80 / 3 tests
    Save £19.20
    5% Off
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Allows quick and easy concentration of antibodies and proteins

The antibody clean up kit method utilizes a simple spin column to easily and quickly remove excess buffer from the antibody thereby providing a more concentrated antibody solution. The clean up kit also allows the experimenter to perform a simple buffer exchange to transfer the antibody into a more favorable conjugation buffer. The kit can also be used to reduce the concentration of many unwanted additives often found in antibody formulations such as azide, glycine or tris.

Recommended Storage

Store at 4°C. Do not freeze.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Antibody Concentration and Clean-up Antibody Purification Kit
Product Line Novus
Content And Storage 4°C. Do not freeze.
Quantity 3 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.