missing translation for 'onlineSavingsMsg'
Learn More

(+)-Abscisic acid, 98%

Product Code. 16338297 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16338297 500 mg 500mg
16328297 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16338297 Supplier Thermo Scientific Acros Supplier No. 342405000
  • ONLINE EXCLUSIVE PRICE
    £176.00
    £158.65 / 500mg
    Save £17.35
    9% Off
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21293-29-8 | C15H20O4 | 264.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21293-29-8
Molecular Formula C15H20O4
Molecular Weight (g/mol) 264.32
MDL Number MFCD00066545,MFCD00066545,MFCD00075619
InChI Key JLIDBLDQVAYHNE-YKALOCIXSA-N
IUPAC Name (2Z,4E)-5-[(1S)-1-hydroxy-2,6,6-trimethyl-4-oxocyclohex-2-en-1-yl]-3-methylpenta-2,4-dienoic acid
SMILES C\C(\C=C\[C@@]1(O)C(C)=CC(=O)CC1(C)C)=C\C(O)=O

Specifications

Melting Point 160.0°C to 163.0°C
Assay Percent Range 98%
Quantity 500 mg
Merck Index 15, 12
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: soluble in aq. nahco3, chloroform, acetone, ethyl, acetate, ether, ethanol, slightly soluble in benzene., sparingly soluble in petroleum ether
Formula Weight 264.32
Chemical Name or Material (+)-Abscisic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.