missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Microscope Slide Storage and Dust Covers
Manufactured from ABS for toughness and impact resistance. Kartell™ Slide Storage and Dust Covers provide a safe and clean environment for microscope slides.
Brand: Kartell™ 0092100
Code : 02
Additional Details : Weight : 0.33652kg
Description
- Accommodates 100 (76L x 26mmW) microscope slides
- Each cavity is numbered; batch data can be written inside or outside of end plates
- Purchase storage units and dust covers separately
- Covers prevent dust and other foreign bodies from contaminating slides
Specifications
100 Place Slide Storage | |
360 x 38 x 100 mm | |
360 mm | |
38 mm |
100 place slide storage unit | |
100 mm | |
ABS, Polystyrene |