missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABRA1 Recombinant Rabbit Monoclonal Antibody (23GB4760), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553840
This item is not returnable.
View return policy
Description
CCDC98, also known as Abraxas 1, was identified through protein binding studies using the breast and ovarian predisposition protein BRCA1 as the binding target. CCDC98 recruits RAP80, a ubiquitin-binding protein, to BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both CCDC98 and RAP80 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either CCDC98 or RAP80 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that CCDC98 and RAP80 control only part of the DNA damage response role of BRCA1. At least two isoforms of CCDC98 are known to exist.
Specifications
| ABRA1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Abraxas1 | |
| 3830405G04Rik; 5630400M01Rik; ABRA1; abraxas protein; Abraxas1; AI506069; AL024423; AV118690; BRCA1-A complex subunit Abraxas; BRCA1-A complex subunit Abraxas 1; Ccdc98; coiled-coil domain containing 98; coiled-coil domain-containing protein 98; FAM175A; family with sequence similarity 175 member A; family with sequence similarity 175, member A; FLJ11520; FLJ12642; FLJ13614; Protein FAM175A; RGD1305287; UNQ496/PRO1013 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84142 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4760 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6UWZ7 | |
| Abraxas1 | |
| A synthesized peptide derived from human CCDC98 (380-409AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction