Learn More
Invitrogen™ ABRA1 Monoclonal Antibody (ABRA1-01)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA110045
Product Code. 12302083
  • £392.00 / 100µg
Description
This antibody recognizes human ABRA1 (Abraxas, CCDC98), an adaptor protein involved in DNA repair, which migrates as a 45 kDa band on PAAGE under reducing conditions.
CCDC98, also known as Abraxas 1, was identified through protein binding studies using the breast and ovarian predisposition protein BRCA1 as the binding target. CCDC98 recruits RAP80, a ubiquitin-binding protein, to BRCA1, allowing the formation of BRCA1 foci in response to DNA damage caused by ionizing radiation. Both CCDC98 and RAP80 are required for DNA damage resistance, G2-M checkpoint control, and DNA repair. Cells depleted of either CCDC98 or RAP80 exhibited increased sensitivity to ionizing radiation, although not as much as in BRCA1-depleted cells, suggesting that CCDC98 and RAP80 control only part of the DNA damage response role of BRCA1. At least two isoforms of CCDC98 are known to exist.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ABRA1
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
Q6UWZ7
Abraxas1
Recombinant protein corresponding to amino acids 1-313 of ABRA1 with N-terminal His6 tag.
100 μg
Primary
Human
Antibody
IgG2a κ
Flow Cytometry, Immunoprecipitation, Western Blot
ABRA1-01
Unconjugated
Abraxas1
3830405G04Rik; 5630400M01Rik; ABRA1; abraxas protein; Abraxas1; AI506069; AL024423; AV118690; BRCA1-A complex subunit Abraxas; BRCA1-A complex subunit Abraxas 1; Ccdc98; coiled-coil domain containing 98; coiled-coil domain-containing protein 98; FAM175A; family with sequence similarity 175 member A; family with sequence similarity 175, member A; FLJ11520; FLJ12642; FLJ13614; Protein FAM175A; RGD1305287; UNQ496/PRO1013
Mouse
Protein A
RUO
84142
4°C, do not freeze