missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527079
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein that is similar to the protein encoded by the breakpoint cluster region gene located on chromosome 22. The protein encoded by this gene contains a GTPase-activating protein domain, a domain found in members of the Rho family of GTP-binding proteins. Functional studies in mice determined that this protein plays a role in vestibular morphogenesis, suggesting that Rho-related GTPases help coordinate motor skills and balance. Alternatively spliced transcript variants that encode different isoforms have been reported for this gene.
Specifications
| ABR | |
| Polyclonal | |
| Unconjugated | |
| ABR | |
| ABR; active BCR-related; active BCR-related gene; active breakpoint cluster region-related protein; MDB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 109934, 29 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q12979, Q5SSL4 | |
| ABR | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ABR. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction