missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563028
This item is not returnable.
View return policy
Description
Immunogen sequence: EESEASPQVH PFPDHELEDM KMKISALKSE IQKEKANKGQ SRAIERLKKK MFENEFLLL Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene encodes a protein that is similar to the protein encoded by the breakpoint cluster region gene located on chromosome 22. The protein encoded by this gene contains a GTPase-activating protein domain, a domain found in members of the Rho family of GTP-binding proteins. Functional studies in mice determined that this protein plays a role in vestibular morphogenesis, suggesting that Rho-related GTPases help coordinate motor skills and balance. Alternatively spliced transcript variants that encode different isoforms have been reported for this gene.
Specifications
| ABR | |
| Polyclonal | |
| Unconjugated | |
| ABR | |
| ABR; active BCR-related; active BCR-related gene; active breakpoint cluster region-related protein; MDB | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q12979 | |
| ABR | |
| Recombinant protein corresponding to Human ABR. Recombinant protein control fragment (Product #RP-101395). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction