missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Abpure™ TCS Antibody Purification Kit

Product Code. 18066404 Shop All Bio Techne Products
Click to view available options
Sufficient For:
1 Purification
3 Purifications
Unit Size:
1 test
3 tests
This item is not returnable. View return policy

Product Code. 18066404

Brand: Novus Biologicals™ 2640030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Novus Biologicals™ Abpure TCS Antibody Purification Kit can be used in its rapid version by spin-purification in centrifuge, or by gravity-flow purification.

  • Fully compatible with InnovaCoat GOLD and LATEX conjugation kits which allow purified antibody to be conjugated rapidly
  • Antibody to be purified should be in 10mL to 50mL of tissue culture supernatant (TCS)
  • Up to 5mg of antibody can be purified in each run
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Quantity 1 Test
Kit Contents 4 or 12 15mL collection tubes, 1 bottle of AbPure elution buffer, 1 bottle of AbPure neutralization buffer, 1 bottle of AbPure preservative, 1 bottle of 10x AbPure binding buffer, 1 bottle of AbPure wash buffer, 1 or 3 bottles of AbPure TCS protein A resi
Sufficient For 1 Purification
Product Type Antibody Purification Kit

For research use only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.