missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABP1/AOC1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15232
This item is not returnable.
View return policy
Description
ABP1/AOC1 Polyclonal specifically detects ABP1/AOC1 in Human samples. It is validated for Western Blot.
Specifications
| ABP1/AOC1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| ABP, amiloride binding protein 1 (amine oxidase (copper-containing)), Amiloride-binding protein, amiloride-sensitive amine oxidase, amiloride-sensitive amine oxidase [copper-containing], AOC1DAOamiloride-binding protein-1, DAO1, Diamine oxidase, EC 1.4.3, EC 1.4.3.22, Histaminase, KAO, Kidney amine oxidase | |
| Rabbit | |
| 85 kDa | |
| 0.1 mL | |
| Immunology | |
| 26 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.55mg/mL | |
| Western Blot 1:500-1:3000 | |
| P19801 | |
| AOC1 | |
| Recombinant protein encompassing a sequence within the center region of human KAO. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction