missing translation for 'onlineSavingsMsg'
Learn More

ABP1/AOC1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18325806 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18325806 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18325806 Supplier Novus Biologicals Supplier No. NBP300162AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ABP1/AOC1 Polyclonal specifically detects ABP1/AOC1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABP1/AOC1
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias ABP, amiloride binding protein 1 (amine oxidase (copper-containing)), Amiloride-binding protein, amiloride-sensitive amine oxidase, amiloride-sensitive amine oxidase [copper-containing], AOC1DAOamiloride-binding protein-1, DAO1, Diamine oxidase, EC 1.4.3, EC 1.4.3.22, Histaminase, KAO, Kidney amine oxidase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant ABP1/AOC1 (AAH14093.1; Met1-Val751).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 26
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.