missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABO Recombinant Rabbit Monoclonal Antibody (23GB3610), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | ABO |
|---|---|
| Clone | 23GB3610 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089923
|
Invitrogen™
MA552620 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089333
|
Invitrogen™
MA552621 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ABO Recombinant Monoclonal antibody specifically detects ABO in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ABO | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P16442, P38649, Q9ET32 | |
| 100301568, 28, 80908 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3610 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| a transferase; A1-specific alpha 1-3-N-acetylgalactosaminyltransferase; A3GALNT; A3GALT1; ABO; ABO A3 transferase; ABO blood group (alpha 1-3-N-acetylgalactosaminyltransferase, alpha 1-3-galactosyltransferase); ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase); ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase; transferase B, alpha 1-3-galactosyltransferase); ABO glycosyltransferase; ABO weak transfer; Abo2; B (A) alpha 1; B blood group galactosyltransferase; b transferase; B(A) alpha-1,3-galactosyltransferase; BGAT; blood group A glycosyltransferase 2; Cis-AB transferase; cis-AB transferase 2; fucosylglycoprotein; fucosylglycoprotein 3-alpha-galactosyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase; Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form; Glycoprotein-fucosylgalactoside alpha-galactosyltransferase; glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase; glycosyltransferase A; glycosyltransferase B; GTB; Histo-blood group A transferase; histo-blood group A2 transferase; histo-blood group ABO system transferase; Histo-blood group ABO system transferase 2; histo-blood group B transferase; NAGAT; NAGAT 2; putative blood group A transferase T2; Transferase; transferase A; transferase B, alpha 1-3-galactosyltransferase) | |
| A synthetic peptide of human BGAT (5-28AA). | |
| Primary | |
| -20°C | |
| ABO |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title