missing translation for 'onlineSavingsMsg'
Learn More

ABO, Blood Group A1B antigen Antibody (HE-24), Novus Biologicals™

Product Code. 18384235 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384235 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384235 Supplier Novus Biologicals Supplier No. NBP311835

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ABO, Blood Group A1B antigen Monoclonal specifically detects ABO, Blood Group A1B antigen in Human samples. It is validated for Flow Cytometry, Agglutination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABO, Blood Group A1B antigen
Applications Flow Cytometry, Agglutination
Classification Monoclonal
Clone HE-24
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1-4 ug/mL, Agglutination
Formulation TBS
Gene Alias A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase;transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Host Species Mouse
Immunogen Glycoprotein fraction isolated from saliva of secretors with blood group A.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.