Learn More
Abnova™ HOXA9/CEN7p (Texas Red/FITC) FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0130.200uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16083195
  • £2030.00 / 200µL
Estimated Shipment: 12-01-2024
to see stock.
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
Store at 4°C in the dark.
Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
7p15.2/7p12.1
HOXA9/CEN7p FISH Probe
Genomic DNA
FITC, Texas Red
DAPI Counterstain (1500 ng/mL ) 250 uL
Approx. 190kb/700kb
200 μL