missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Fluorescent In Situ Hybridization (FISH) Buffer

Product Code. p-4370464
Click to view available options
Quantity:
1 mL
5 x 1 mL
Unit Size:
1mL
Pack of 5
This item is not returnable. View return policy

Product Code. 16082368

Brand: Abnova™ U0028.1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimized for performance with FISH probes

Fluorescent In Situ Hybridization (Cell)

Specifications

Content And Storage Store at 4°C
Description FISH Hybridization Buffer is optimized for performance with FISH probes and contains dextran sulfate, formamide, and SSC (pH 7.0).
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Product Type FISH Hybridization Buffer(1ml)
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.