missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova ES-D3 (Mouse pluripotent embryonic stem cell) whole cell lysate, nondenatured
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Beskrivning
- Stem cells maintained in the undifferentiated state using specialized culturing protocols
- Monitored for undifferentiated cell morphology
- Analyzed using Flow Cytometry against specific cell marker
- Coommasie Blue Staining of total cell lysate in SDS-PAGE
Western Blotting, Immunoprecipitation
Specifikationer
Specifikationer
| For Use With (Application) | Immunoprecipitation, Western Blot |
| Tissue | Embryonic |
| Description | Whole cell lysate (non-denatured) |
| Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Preparation Method | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). |
| Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Quantity | 50 μg |
| Storage Buffer | In modified RIPA Lysis Buffer. |
| Host Species | Mouse |
| Concentration | 4 mg/mL |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering