missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CEN4q (FITC) FISH Probe

Product Code. 16082605
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
This item is not returnable. View return policy

Product Code. 16082605

Brand: Abnova™ FC0031.20uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

FISH Hybridization Buffer (80μL)

Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.

FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.

Advantages

  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity

Specifications

Content And Storage Store at 4°C in the dark.
Label or Dye FITC
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Includes FISH Hybridization Buffer (80 uL)
Location 4q12
Probe CEN4q
Product Type CEN4q (FITC) FISH Probe
Quantity 20 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.