missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CCND1 (Orange) FISH Probe
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
- Dual colored probes for fast, sensitive, and specific detection
- Work on metaphase spread, paraffin embedded, and frozen tissue
- Identify gene amplification, loss, and translocation
- High signal to noise ratio
- Low cross reactivity
Specifications
Specifications
| Content And Storage | Store at -20°C in the dark. |
| Description | Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique. |
| Label or Dye | Orange |
| For Use With (Application) | Fluorescent In Situ Hybridization |
| Location | 11q13 |
| Probe | Approx. 162kb |
| Product Type | CCND1 (Orange) FISH Probe |
| Quantity | 200 μL |
| Source | Genomic DNA |
| Species | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction