missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABLIM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551874
This item is not returnable.
View return policy
Description
Immunogen sequence: DLSTATKSKT SEDISQTSKY SPIYSPDPYY ASESEYWTYH GSPKVPRARR FSSGGEEDDF DRSMHKLQSG IGRLILKEEM KARSSSYADP WTPPRSSTSS REALHTAGYE MSLNGSPRSH YLADSDPLIS KSASLPAYRR NGLHRT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
The LIM domain is a double zinc finger structure that promotes protein-protein interactions. LIM domain proteins, such as ABLIM3, play roles in embryonic development, cell lineage determination, and cancer (Krupp et al., 2006 ).
Specifications
| ABLIM3 | |
| Polyclonal | |
| Unconjugated | |
| Ablim3 | |
| ABLIM3; abLIM-3; actin binding LIM protein family member 3; actin binding LIM protein family, member 3; actin-binding LIM protein 3; Actin-binding LIM protein family member 3; AW990719; D930036B08Rik; HMFN1661; KIAA0843; RGD1565118 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22885 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94929 | |
| Ablim3 | |
| Recombinant protein corresponding to Human ABLIM3. Recombinant protein control fragment (Product #RP-102137). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction