missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABLIM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557405
This item is not returnable.
View return policy
Description
Immunogen sequence: DGEDGSLDQD NRKKSSWLML KGDADTRTNS PDLDTQSLSH SSGTDRDPLQ RMAGDSFHSR FPYSKSDPLP GHGKNGLDQR NANLAPCGAD PDAS Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 78%.
ABLIM2 (Actin Binding LIM Protein Family Member 2) is a Protein Coding gene. Among its related pathways areDevelopmental BiologyandAxon guidance. Gene Ontology (GO) annotations related to this gene includeactin binding. An important paralog of this gene isABLIM1.
Specifications
| ABLIM2 | |
| Polyclonal | |
| Unconjugated | |
| Ablim2 | |
| Ablim2; abLIM-2; actin binding LIM protein 2; actin binding LIM protein family member 2; actin binding LIM protein family, member 2; actin-binding LIM protein 2; Actin-binding LIM protein family member 2; AI606905; C230091L11; KIAA1808; testis tissue sperm-binding protein Li 35a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84448 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6H8Q1 | |
| Ablim2 | |
| Recombinant protein corresponding to Human ABLIM2. Recombinant protein control fragment (Product #RP-96714). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction