missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABLIM1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99386CL1
This item is not returnable.
View return policy
Description
ABLIM1 Polyclonal antibody specifically detects ABLIM1 in Human samples. It is validated for Western Blot
Specifications
| ABLIM1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3983 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Western Blot | |
| CoraFluor 1 | |
| ABLIM, abLIM-1, actin binding LIM protein 1, Actin-binding double zinc finger protein, actin-binding double-zinc-finger protein, actin-binding LIM protein 1, Actin-binding LIM protein family member 1, DKFZp781D0148, FLJ14564, KIAA0059, LIM actin-binding protein 1, LIMAB1MGC1224, LIMATIN | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ABLIM1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction