missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABL2 (Arg) Recombinant Human Protein
FLT1 (VEGFR1) Recombinant Human Protein
£438.00 - £6900.00
Specifications
Accession Number | NP_009298.1 |
---|---|
Molecular Weight (g/mol) | 130.9kDa |
Name | ABL2 (Arg) Recombinant Human Protein |
Storage Requirements | –80°C |
Regulatory Status | For research use only. Not for use in diagnostic procedures |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11553407
|
Invitrogen
PV3266 |
10μg |
£438.00
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10104852
|
Invitrogen
PV3586 |
1mg |
£6900.00
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ABL2 (Arg) is an Abl-related non-receptor protein tyrosine kinase that links cell surface receptors to cell proliferation and⁄or cytoskeletal reorganization.Specifications
NP_009298.1 | |
ABL2 (Arg) Recombinant Human Protein | |
For research use only. Not for use in diagnostic procedures | |
ABL2 (Arg) | |
ABL2 (ARG) was pre-diluted in enzyme dilution buffer and assayed in 50mM Tris (pH 7.5), 10mM MgCl2, 1mM DTT, 0.01% NP-40, 200μM ATP, 100μM Abl1 peptide substrate (EAIYAAPFAKKK) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
ABL2 (Arg) Recombinant Human Protein | |
His Tag |
130.9kDa | |
–80°C | |
1 Each | |
ABL2 (Arg) Recombinant Human Protein | |
ABL | |
Human | |
Baculovirus |
For Research Use Only. Not for use in diagnostic procedures.